Learn More
Honeywell Salisbury™ External Rod Clampsticks
Easy-grip plastic hand grip
Supplier: Honeywell Salisbury™ 4011
Description
- Constructed of closed-cell foam-filled tubular fiberglass in accordance with ASTM F711
- Hook operating end made of solid 3/8 in. (9.5mm) dia. fiberglass and mounted on tool exterior for easy wipedown
- Thermoplastic head ferrule, stainless-steel hook, heavy rubber end cap
Specifications
External Rod Clampstick | |
1.25 in. | |
10.8 x 120 x 8 in. (320 x 304 x 20.3 cm) | |
20.3 cm | |
10.8 ft. | |
7 lb. | |
120 in. |
Foam-filled Fiberglass | |
3.17 cm | |
8 in. | |
Tough Thermoplastic head ferrule, EZ Grip Plastic hand grip, Stainless Steel hook, Heavy duty Rubber end cap | |
320 cm | |
3.2 kg | |
304 cm |