Learn More
Honeywell Salisbury™ External Rod Clampsticks
Easy-grip plastic hand grip
Supplier: Honeywell Salisbury™ 4013
Description
- Constructed of closed-cell foam-filled tubular fiberglass in accordance with ASTM F711
- Hook operating end made of solid 3/8 in. (9.5mm) dia. fiberglass and mounted on tool exterior for easy wipedown
- Thermoplastic head ferrule, stainless-steel hook, heavy rubber end cap
Specifications
External Rod Clampstick | |
1.25 in. | |
14.8 x 168 x 8 in. (440 x 426 x 20.3 cm) | |
20.3 cm | |
14.8 ft. | |
9.4 lb. | |
168 in. |
Foam-filled Fiberglass | |
3.17 cm | |
8 in. | |
Tough Thermoplastic head ferrule, EZ Grip Plastic hand grip, Stainless Steel hook, Heavy duty Rubber end cap | |
440 cm | |
4.3 kg | |
426 cm |