Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ LP Series High Voltage Goatskin Leather Lineman's Glove Protectors

Catalog No. 19828376 Shop All WH Salisbury & Co. Products
Change view
Click to view available options
Size:
7
8
9
10
11
12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-828-376 8
19-828-375 7
19-828-377 9
19-828-378 10
19-828-379 11
19-828-380 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-828-376 Supplier Honeywell Salisbury™ Supplier No. ILPG3S/8
This item is not returnable. View return policy

Glove protectors are 12 in. long and feature adjustable straps with non-metallic buckles. Wear over insulating rubber gloves for mechanical protection against cuts, abrasions, and punctures.

  • Constructed of high quality, top grain goatskin leather
  • Adjustable straps with non-metallic buckles
  • Wear over insulating rubber gloves for mechanical protection
  • Ideal for industrial electrical workers in high voltage situations
TRUSTED_SUSTAINABILITY

Specifications

Back Material Vinyl
Certifications/Compliance Meets ASTM F696 Specifications
Applications Industrial
Color Cream
Material Top Grain Goatskin Leather
Product Type Glove Protectors
Size 8
For Use With (Application) Mechanical Protection
For Use With (Equipment) Rubber Insulating Gloves
Glove Cuff Color Orange
Glove Cuff Style Gauntlet
Series ILPG3S
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.