Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Salisbury™ Grain Cowhide Leather Linesmen's Work Gloves
Shop All WH Salisbury & Co. Products
Click to view available options
Size:
Small
X-Large
Description
- Designed specifically for use by linemen
- Gloves offer many of the quality features found in leather protectors
- Grain cowhide leather is soft and flexible, while still being highly resistant to cuts and abrasions
- Natural pigskin leather cuff
Specifications
Specifications
| Cuff Material | Pigskin Leather |
| Color | Yellow |
| Hand | Specific |
| Material | Grain Cowhide Leather |
| Size | Small |
| For Use With (Application) | Linesmen's Work |
| Glove Cuff Color | Brown |
| Glove Cuff Style | Gauntlet |
| Series | LW2SPE |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction