Learn More
Honeywell Salisbury™ Pro-Wear™ Arc Flash Protection Jacket Kits
Catalog No. p-2816483 Shop All WH Salisbury & Co. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
11cal/cm2 Flash Kit without gloves
Kits contain an arc flash jacket with hood, an AS1000HAT, SKBAG and safety glasses.
  • ATPV rating: 12cal/cm2
  • Jackets are made from arc flash resistant, Indura Ultra Soft™
  • Sewn with Nomex™ thread
  • Jacket length: 32 in. long
  • Nomex wristlets; hook and pile front closure
Order Info
Insulating rubber gloves are not included with these kits.
Certifications
Hazard Risk Category 2. CE marked, CE 0120 certified.
Compliance
Kits meet NFPA 70E-2009 and ASTM™ F1506 standards.
TRUSTED_SUSTAINABILITY
Specifications
Arc Flash Rating 11 cal/cm²
Description Coat, bib overalls, Pro-Hood™, hard hat, Salisbury storage bag and safety Glasses
Certifications/Compliance NFPA 70E™ 2009, CE 0120
Size Large
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.