Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SALL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552065 Supplier Invitrogen™ Supplier No. PA552065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLAHQNACST DPPVMVIIGG QENPNNSSAS SEPRPEGHNN PQVMDTEHSN PPDSGSSVPT DPTWGPERRG EESSGHFLVA ATGTAAGGGG GLILASPKLG ATPLPPESTP A Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

This gene encodes a protein containing multiple zinc finger domains. The encoded protein functions in optical fissure closure during development of the eye in the embryo. Mutations in this gene are associated with ocular coloboma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SALL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SALL2
Gene Accession No. Q9Y467
Gene Alias AI225809; AW559097; COLB; HSAL2; KIAA0360; mKIAA0360; Msal-2; p150(Sal2); SAL2; Sal-2; SALL2; sal-like 2; sal-like 2 (Drosophila); sal-like protein 2; spalt like transcription factor 2; spalt-like protein 2; spalt-like transcription factor 2; zinc finger protein 795; Zinc finger protein SALL2; zinc finger protein Spalt-2; ZNF795
Gene Symbols SALL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SALL2. Recombinant protein control fragment (Product #RP-89065).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6297
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.