Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SALL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571337 Supplier Invitrogen™ Supplier No. PA571337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SALL2 Polyclonal Antibody for Western Blot

This gene encodes a protein containing multiple zinc finger domains. The encoded protein functions in optical fissure closure during development of the eye in the embryo. Mutations in this gene are associated with ocular coloboma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SALL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SALL2
Gene Accession No. Q9Y467
Gene Alias AI225809; AW559097; COLB; HSAL2; KIAA0360; mKIAA0360; Msal-2; p150(Sal2); SAL2; Sal-2; SALL2; sal-like 2; sal-like 2 (Drosophila); sal-like protein 2; spalt like transcription factor 2; spalt-like protein 2; spalt-like transcription factor 2; zinc finger protein 795; Zinc finger protein SALL2; zinc finger protein Spalt-2; ZNF795
Gene Symbols SALL2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SALL2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6297
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.