Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SALL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540564
Description
Peptide sequence: SVENGSGESD RLSNDSSSAV GDLESRSAGS PALSESSSSQ ALSPAPSNGE Sequence homology: Cow: 79%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
SALL3 belongs to the sal C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. SALL3 is a probable transcription factor.
Specifications
SALL3 | |
Polyclonal | |
Unconjugated | |
SALL3 | |
B130022O04Rik; C2H2 zinc finger protein SALL3; hSALL3; MSal; Msal-1; Sal; Sall3; sal-like 3; sal-like 3 (Drosophila); sal-like protein 3; Salt; Spalt; spalt like transcription factor 3; spalt-like protein 3; spalt-like transcription factor 3; spalt-like zinc finger protein; zinc finger protein 796; Zinc finger protein SALL3; ZNF796 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27164 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BXA9 | |
SALL3 | |
Synthetic peptide directed towards the middle region of human SALL3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction