Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SALL4 Antibody (CL12183), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321209100UL
Description
SALL4 Monoclonal antibody specifically detects SALL4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SALL4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
57167 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CL12183 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
dJ1112F19.1, DRRS, HSAL4, MGC133050, sal-like 4 (Drosophila), sal-like protein 4, Zinc finger protein 797, Zinc finger protein SALL4, ZNF797sal (Drosophila)-like 4 | |
This antibody was developed against a Synthetic Peptide of Q9UJQ4. Actual immunogen sequence is proprietary. | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction