Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SALL4 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298704JF549
Description
SALL4 Polyclonal specifically detects SALL4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SALL4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
dJ1112F19.1, DRRS, HSAL4, MGC133050, sal-like 4 (Drosophila), sal-like protein 4, Zinc finger protein 797, Zinc finger protein SALL4, ZNF797sal (Drosophila)-like 4 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SALL4. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
57167 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction