Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SALL4 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993000
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3000 10 μL
50-199-2999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3000 Supplier Active Motif Supplier No. 39958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sall4 protein is encoded by the Sall4 gene located on chromosome 20 in humans and belongs to the Spalt-like protein family. It plays an important role in somatic stem cell maintenance and is studied as a stem cell biomarker. It is a zinc finger transcription factor that is upregulated during development and shown to interact with Oct4 and Sox2. It also forms a complex with Nanog to maintain the pluripotency of Embryonic Stem cells (ESCs). Several current research studies focus on its function as an oncogene and tumor biomarker. Some of the reported target genes of Sall4 include p53, Bmi-1, Bcl2 and XIAP.

Specifications

Antigen SALL4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene SALL4
Gene Accession No. Q8BX22
Gene Alias 5730441M18Rik; AA407717; AL022809; AW536104; C330011P20Rik; C78083; C78563; CH242-152A8.2; dJ1112F19.1; DRRS; HSAL4; SALL4; sal-like 4; sal-like 4 (Drosophila); Sal-like protein 4; spalt like transcription factor 4; spalt-like transcription factor 4; Tex20; Zinc finger protein 797; Zinc finger protein SALL4; ZNF797
Gene Symbols SALL4
Host Species Rabbit
Immunogen This SALL4 antibody was raised against a synthetic peptide derived from the N-terminus of human SALL4.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 99377
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.