Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SALL4 Rabbit anti-Human, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501734392 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4392 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4392 Supplier Proteintech Group Inc Supplier No. 245001AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sall4 is a probable transcription factor. Defects in Sall4 are the cause of Okihiro syndrome; also known as Duane radial ray syndrome (DRRS). It is a disorder characterized by the association of forearm malformations with Duane retraction syndrome. Sall4 is also involved in forelimb and heart development in mice.

Specifications

Antigen SALL4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SALL4
Gene Accession No. Q9UJQ4
Gene Alias DRRS, HSAL4, sal like 4 (Drosophila), Sal like protein 4, SALL4, Zinc finger protein SALL4, ZNF797
Gene Symbols SALL4
Host Species Rabbit
Immunogen SALL4 Fusion Protein Ag17480
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57167, 686412
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.