Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella Antibody (100/358) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064563
Description
Salmonella Monoclonal specifically detects Salmonella in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Salmonella | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Native S. enteritidis (NCTC 5188 9.12:g,m). | |
0.1 mg | |
Primary | |
Bacteria | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
100/358 | |
Unconjugated | |
Mouse | |
Protein A purified | |
RUO | |
Antibody recognizes an antigen common to the various Salmonella strains. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction