Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salmonella enteritidis H Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988698
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8698 Supplier Bioss Supplier No. BS8670R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHP-1 (PTPN6) is a non-receptor protein tyrosine phosphatase that is expressed primarily in hematopoietic cells. The enzyme is composed of two SH2 domains, a tyrosine phosphatase catalytic domain, and a carboxy-terminal regulatory domain. SHP-1 removes phosphates from target proteins to downregulate several tyrosine kinase-regulated pathways. In hematopoietic cells, the amino-terminal SH2 domain of SHP-1 binds to tyrosine phosphorylated erythropoietin receptors (EpoR) to negatively regulate hematopoietic growth. Overexpression of SHP-1 in epithelial cells results in dephosphorylation of the Ros receptor tyrosine kinase and subsequent downregulation of Ros-dependent cell proliferation and transformation. Following ligand binding in myeloid cells, SHP-1 associates with the IL-3R beta chain and downregulates IL-3-induced tyrosine phosphorylation and cell proliferation.

Specifications

Antigen Salmonella enteritidis H
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTPN6
Gene Accession No. P29350
Gene Alias 70Z-SHP; EC 3.1.3.48; HCP; Hcph; hematopoietic cell phosphatase; Hematopoietic cell protein-tyrosine phosphatase; HPTP1C; me; motheaten; protein tyrosine phosphatase, non-receptor type 6; Protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTN6; Ptp1C; PTP-1C; Ptph6; PTPN6; PTPTY-42; SH2 phosphatase 1; SHP1; SHP-1; SHP-1L; SH-PTP1; Tyrosine-protein phosphatase non-receptor type 6
Gene Symbols PTPN6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Salmonella enteritidis H.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5777
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.