Learn More
Invitrogen™ Salmonella LPS Monoclonal Antibody (D46J)

Description
This antibody is specific for common LPS core of all Salmonellae O-serogroups tested: A, B, C1, C2, D, E1, E3, E4, F, G1 and G2. This does not cross-react with E. coli (mix), Klebsiella, Citrobacter, Pseudomonas, Yersinia, Shigella, Proteus or Legionella. Purity is >90%. Recommended pairs for ELISA: MA5-18257 (capture) and MA5-18259 or MA5-18260 (conjugated, detection).
Specifications
Specifications
| Antigen | Salmonella LPS |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | D46J |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Salmonellae |
| Host Species | Mouse |
| Immunogen | Salmonellae sp. native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.