Learn More
Invitrogen™ Salmonella paratyphi A Monoclonal Antibody (10B10G)

Description
This antibody detects Salmonella paratyphi. It does not react with other Salmonella, E. coli, or Klebsiella.
Specifications
Specifications
| Antigen | Salmonella paratyphi A |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 10B10G |
| Concentration | 6.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | Salmonellae |
| Host Species | Mouse |
| Immunogen | Heat inactivated bacterial cells of Salmonella paratyphi A. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.