Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Salmonella paratyphi C2 LPS Monoclonal Antibody (6348)
GREENER_CHOICE

Catalog No. PIMA533294
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533294 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533294 Supplier Invitrogen™ Supplier No. MA533294
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Salmonella paratyphi C2 LPS Monoclonal Antibody for ICC/IF, ELISA

Salmonella paratyphi C2 LPS (lipopolysaccharide) is a complex molecule found on the outer membrane of the bacterium Salmonella paratyphi C2. It is composed of lipid A, core oligosaccharide, and O-antigen polysaccharide chains. The O-antigen polysaccharide chains are highly variable and are responsible for the serological diversity of Salmonella strains. The LPS of Salmonella paratyphi C2 is of particular interest as it is associated with the pathogenesis of typhoid fever, a serious and potentially fatal disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Salmonella paratyphi C2 LPS
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6348
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Salmonellae
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.