Learn More
Invitrogen™ Salmonella Polyclonal Antibody, Biotin

Description
The PA1-73022 antibody reacts with Salmonella species. PA1-73022 has been successfully used in Immunofluorescence and ELISA applications. The PA1-73022 antibody was raised against a mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens). Recommended short term storage, up to 6 months, at 4°C. For longer term storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Salmonella |
| Applications | ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Salmonellae |
| Host Species | Rabbit |
| Immunogen | Mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens). |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.