Learn More
Invitrogen™ Salmonella Polyclonal Antibody, FITC

Description
The PA1-73020 antibody reacts with Salmonella species and crossreacts with related enterobacteriaceae. PA1-73020 has been successfully used in Immunofluorescence applications. Acetone fixation of the antigen source is recommended prior to staining. The PA1-73020 antibody was raised against a mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens).
Specifications
Specifications
| Antigen | Salmonella |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | Salmonellae |
| Host Species | Rabbit |
| Immunogen | Mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens). |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.