Learn More
Invitrogen™ Salmonella Polyclonal Antibody, HRP

Description
The PA1-73021 antibody reacts with Salmonella species. PA1-73021 has been successfully used in ELISA applications. The PA1-73021 antibody was raised against a mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens). Recommended short term storage, up to 6 months, store at 4°C. For longer term storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Salmonella |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2 |
| Gene Alias | Salmonellae |
| Host Species | Rabbit |
| Immunogen | Mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens). |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.