Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Salmonella typhi hupB Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144578 Supplier Invitrogen™ Supplier No. PA5144578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Salmonella typhi hupB Polyclonal Antibody for Western Blot, ELISA

Histone-like DNA-binding protein which is capable of wrapping DNA to stabilize it, and thus to prevent its denaturation under extreme environmental conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Salmonella typhi hupB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene hupB
Gene Accession No. P0A1R9, P64389
Gene Alias DNA-binding protein HU-beta; hupB
Gene Symbols hupB
Host Species Rabbit
Immunogen Recombinant Neisseria meningitidis serogroup B hupB protein (1-89aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57493645, 61281390
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.