Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Salmonella typhi hupB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117630
Description
This antibody has been tested in direct-ELISA.
Histone-like DNA-binding protein which is capable of wrapping DNA to stabilize it, and thus to prevent its denaturation under extreme environmental conditions.
Specifications
Salmonella typhi hupB | |
Polyclonal | |
Unconjugated | |
hupB | |
DNA-binding protein HU-beta; hupB | |
Rabbit | |
Protein G | |
RUO | |
57493645, 61281390 | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P0A1R9, P64389 | |
hupB | |
Recombinant Salmonella typhi DNA-binding protein HU-beta protein (1-90aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction