Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Salmonella typhi Monoclonal Antibody (AC04)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA546571
This item is not returnable.
View return policy
Description
Salmonella typhi Monoclonal Antibody for ICC/IF, ELISA
A ubiquitous human and animal pathogen, salmonella is the causative agent of salmonellosis, a disease which is common throughout the world. The disease is usually not severe in humans, but can occasionally cause serious systemic infections which require antibiotic treatment. As salmonellosis is a zoonosis with a large animal reservoir, contaminated food is by far the most common mode of transmission of non-typhoidal salmonellae. The large and varied animal reservoir of salmonella has made control of salmonellosis a difficult task. While an effective vaccine does exist against typhoid fever, there is none against non-typhoidal salmonellosis.
Specifications
Salmonella typhi | |
Monoclonal | |
0.3 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
AC04 | |
Unconjugated | |
S. typhi | |
Native Salmonella typhi. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction