Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Salmonella typhi Monoclonal Antibody (AC04)
GREENER_CHOICE

Catalog No. PIMA546572
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546572 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546572 Supplier Invitrogen™ Supplier No. MA546572
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Salmonella typhi Monoclonal Antibody for ICC/IF, ELISA

A ubiquitous human and animal pathogen, salmonella is the causative agent of salmonellosis, a disease which is common throughout the world. The disease is usually not severe in humans, but can occasionally cause serious systemic infections which require antibiotic treatment. As salmonellosis is a zoonosis with a large animal reservoir, contaminated food is by far the most common mode of transmission of non-typhoidal salmonellae. The large and varied animal reservoir of salmonella has made control of salmonellosis a difficult task. While an effective vaccine does exist against typhoid fever, there is none against non-typhoidal salmonellosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Salmonella typhi
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone AC04
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias S. typhi
Host Species Mouse
Immunogen Native Salmonella typhi.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.