Learn More
Salmonella typhi Mouse anti-All, Unconjugated, Clone: Clone 4, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Salmonella typhi |
| Applications | ELISA, Immunodiffusion, Western Blot |
| Classification | Monoclonal |
| Clone | Clone 4 |
| Conjugate | Unconjugated |
| Gene Alias | Anti-S. typhi |
| Host Species | Mouse |
| Immunogen | Negative S. typhi Antigen |
| Purification Method | Affinity Purified |
| Quantity | 10 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.