Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Salmonella typhimurium LPS Monoclonal Antibody (1E6)
GREENER_CHOICE

Catalog No. MA122693
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA122693 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA122693 Supplier Invitrogen™ Supplier No. MA122693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-22693 detects Salmonella typhimurium 0 - 4 in bacteria samples.

The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Salmonella typhimurium LPS
Applications Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1E6
Concentration 4.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias S. typhimurium; Salmonella LPS; Salmonellae
Host Species Mouse
Immunogen Lipopolysaccharides (LPS) of S. typhimurium.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.