Learn More
Sigma Aldrich SAM Formation reagent for carboxylic acid terminated monolayer on gold surface
Catalog No. 7466141SET Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 sEt
Catalog No. 74-661-41SET
Supplier: Sigma Aldrich 7466141SET
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Carboxylic acid- SAM is an acid terminated self-assembled monolayer on the surface of gold, which is mainly utilized to immobilize the surface atoms. The SAM film formed has a high degree of molecular orientation. This reagent is majorly utilized in surface science, biomineralization, and sensor based applications.Used to prepare carboxylic acid- terminated self-assembled monolayers (SAMs) on gold surfaces as biosensors detected by QCM (Quartz Crystal Microbalance), SPR (Surface Plasmon Resonance), and electrochemical analysis.Coupled with carboxyl group activation method using NHS/WSC, carboxylic acid-SAM can be used as an interface to immobilize proteins or peptides and other ligands on gold surfaces.This SAM Formation Reagent presents less non-specific binding than those commonly prepared with carboxylic acid-SAM.Storage: Keep tightly closed; packed under Nitrogen gas; store in cooler (0∼5°C); dark and dry place.
Specifications
Quantity 1 sEt
Synonym Carboxylic acid-SAM
Biological Activity Respiratory system
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.