Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAM68 Antibody (SR2119), Novus Biologicals™
SDP

Catalog No. NB258368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258368 100 μg
NB258261 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258368 Supplier Novus Biologicals Supplier No. NBP322041100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SAM68 Monoclonal antibody specifically detects SAM68 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen SAM68
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR2119
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias FLJ34027, GAP-associated tyrosine phosphoprotein p62, KH domain containing, RNA binding, signal transduction associated 1, KH domain-containing, RNA-binding, signal transduction-associated protein 1, p21 Ras GTPase-activating protein-associated p62, p62, p68, SAM68, Sam68GAP-associated tyrosine phosphoprotein p62 (Sam68), Src-associated in mitosis 68 kDa protein
Host Species Rabbit
Immunogen A synthesized peptide derived from human SAM68 (Uniprot #: Q07666)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10657
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.