Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAM68 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118016
This item is not returnable.
View return policy
Description
SAM68 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
SAM68 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q07666 | |
KHDRBS1 | |
E. coli-derived Human SAM68 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
KHDRBS1 | |
FLJ34027; GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KH RNA binding domain containing, signal transduction associated 1; KHDRBS1; khdrbs1 {ECO:0000312; p21 Ras GTPase-activating protein-associated p62; P62; p68; RGD:621459}; Sam68; src associated in mitosis, 68 kDa; src-associated in mitosis 68 kDa protein | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10657 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction