Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAM68 Recombinant Rabbit Monoclonal Antibody (JE47-15)
GREENER_CHOICE

Catalog No. PIMA534847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534847 Supplier Invitrogen™ Supplier No. MA534847
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: SHSY-5Y, mouse brain tissue, A431, F9, HepG2, human liver cancer tissue, human colon cancer tissue, human stomach cancer tissue.

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAM68
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE47-15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene KHDRBS1
Gene Accession No. Q07666
Gene Alias FLJ34027; GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KH RNA binding domain containing, signal transduction associated 1; KHDRBS1; khdrbs1 {ECO:0000312; p21 Ras GTPase-activating protein-associated p62; P62; p68; RGD:621459}; Sam68; src associated in mitosis, 68 kDa; src-associated in mitosis 68 kDa protein
Gene Symbols KHDRBS1
Host Species Rabbit
Immunogen Recombinant protein within Human SAM68 aa 1-108.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10657
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.