Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium cobalt, REacton™, Sm 33%

Catalog No. p-7052180
Click to view available options
Quantity:
50 g
250 g

CAS: 12017-68-4 | CoSm | 209.293 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12017-68-4
Molecular Formula CoSm
Molecular Weight (g/mol) 209.293
MDL Number MFCD02091682
InChI Key KPLQYGBQNPPQGA-UHFFFAOYSA-N
Synonym samarium cobalt, samarium cobalt powder,-30 mesh, cobalt-samarium, samarium-cobalt, powder30mesh, smco5, acmc-20ajrh, cobalt samarium, samarium cobalt, powder, sm2co17
PubChem CID 10125161
IUPAC Name cobalt;samarium
SMILES [Co].[Sm]

Specifications

Physical Form Fine Powder
Quantity 50 g
UN Number UN3089
Grade Reacton™
Odor Odorless
Chemical Name or Material Samarium cobalt, Intermetallic, Magnet Alloy Additive
Hazard Category H228-H317-H334-H350
Hazard Statement GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P240-P241-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 234-625-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.