Alfa Aesar™ Samarium(II) iodide, 0.07 - 0.12M in THF, stab.

Manufacturer: Alfa Aesar™ 22360AE

Quantity 100mL

 View more versions of this product

Catalog No. AA22360AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32248-43-4
    Formula Weight 404.21
    MDL Number MFCD00058873
    UN Number UN3271
    Chemical Name or Material Samarium(II) iodide, Stabilized

    Specifications

    Formula Weight 404.21
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H224-H260-H314-H318
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 0.922g/mL
    Physical Form Liquid
    UN Number UN3271
    Health Hazard 1 Danger
    Concentration 0.07 to 0.12M in THF
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00058873
    CAS 32248-43-4
    Quantity 100mL
    Odor Ether-like
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHERS, N.O.S.
    Chemical Name or Material Samarium(II) iodide, Stabilized

    GHS H Statement
    H224-H260-H314-H318
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit