Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium(III) oxide, 99.9% (metals basis)

Catalog No. AA3627818
Click to view available options
Quantity:
10 g
50 g

CAS: 12060-58-1 | Sm2O3

The biocompatibility of natural samarium (III) oxide, which has previously been used for treatment in bone-related diseases was determined as a first step in its evaluation as a bone implant material. The rates and products of the purely heterogeneous oxidations of C2H6 (g) and C2H4 (g) on Sm2O3 in the presence of O2 (g) were investigated in a very low-pressure flow reactor by on-line molecular beam mass spectrometry, about 1000±100 K. Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. Also, it is used as a neutron absorber in control rods for nuclear power reactors. The oxide catalyzes dehydration of acyclic primary alcohols to aldehydes and ketones. Another use involves preparation of other samarium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-58-1
Molecular Formula Sm2O3
MDL Number MFCD00011237

Specifications

Melting Point 2300°C
Color White
Physical Form Sintered Lump
Quantity 50 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Insoluble in water
Formula Weight 348.7
Refractive Index 1.97
Appearance Off-white 3-12mm Sintered Lump
Chemical Name or Material Samarium(III) oxide
EINECSNumber 235-043-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.