Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium(III) trifluoromethanesulfonate, 98%

Catalog No. AAL2025314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2025314 25 g
AAL2025306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2025314 Supplier Thermo Scientific Chemicals Supplier No. L2025314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 52093-28-4 | C3F9O9S3Sm | 597.55 g/mol

Used as catalyst for Baylis-Hillman reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as catalyst for Baylis-Hillman reaction.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-28-4
Molecular Formula C3F9O9S3Sm
Molecular Weight (g/mol) 597.55
MDL Number MFCD00192431
InChI Key DDCWGUIPLGMBPO-UHFFFAOYSA-K
Synonym samarium iii trifluoromethanesulfonate, samarium triflate, samarium iii triflate, samarium trifluoromethanesulfonate, trifluoromethanesulfonic acid samarium iii salt, samarium iii trifluoromethanesulfonate 97+%, sm otf 3, 3cf3o3s.sm, samarium 3+ tritriflate
PubChem CID 10145316
IUPAC Name samarium(3+);trifluoromethanesulfonate
SMILES [Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 597.56
Percent Purity 98%
Chemical Name or Material Samarium(III) trifluoromethanesulfonate
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.