Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ UrineGARD™ Urine Collection Container

Catalog No. 13303001
Click to view available options
Volume (Metric):
2.5 L
3.5 L

Prevent light exposure using tinted, Thermo Scientific™ Samco™ UrineGUARD™ Urine Collection Containers, which alleviate mess and exposure in urine collection and sampling.

Prevent light exposure by using Thermo Scientific™ Samco™ UrineGUARD™ Urine Collection Containers. The Samco 24-hour Urine Collection product line alleviates mess and exposure when handling urine collection and sampling. Containers are inert to normal preservative.

  • 100% High Density Polyethylene (HDPE)
  • Large carrying handles for easy transportation
  • Easy-to-read graduations and highly accurate scales
  • Custom cap and mouth design helps prevent leakage
  • Upright configuration for compact storage
  • Custom spout allows for accurate pouring into test tubes, centrifuge tubes or aliquot bottles
TRUSTED_SUSTAINABILITY

Specifications

Product Type Urine Collection Container
Closure Type Wide Mouth Screw Cap with Integral Spout
Graduated Yes
Material HDPE
Color Amber
Description 24-hour UrineGUARD Specimen Container
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
No. per Case 24/Cs.
Volume (Metric) 3.5 L
Green Features Less waste
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.