Learn More
Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers

Description
Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:
- Bio-Tite caps create leak-resistant seal for superior safety and handling
- 90mL (3 oz.); Opening: 53mm
- Dual-threaded caps create improved sealing
- Sterilized by gamma irradiation, which eliminates potential contamination
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Stackable containers are easy to store, saving lab space
- Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Material | Virgin Polypropylene |
| Shape | Cylindrical |
| Sterility | Sterile |
| Capacity (Metric) | 90 mL |
| Closure Color | Orange |
| Closure Size | 53 mm |
| Closure Type | Bio-Tite Lid |
| Description | Cap and container assembled |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.