Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers
GREENER_CHOICE

Catalog No. p-4525852
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Natural
Orange
White
Yellow
Label:
Non-tabbed
Tabbed
Tabbed Temp Strip
No. per Case:
300/Cs., Pre-assembled
400/Cs., Pre-assembled
This item is not returnable. View return policy
This item is not returnable. View return policy

Prevent bio-hazardous spills with these leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.

Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:

  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • 90mL (3 oz.); Opening: 53mm
  • Dual-threaded caps create improved sealing
  • Sterilized by gamma irradiation, which eliminates potential contamination
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Stackable containers are easy to store, saving lab space
  • Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage
TRUSTED_SUSTAINABILITY

Specifications

Product Type Specimen Container
Closure Included Yes
Material Virgin Polypropylene
Shape Cylindrical
Sterility Sterile
Capacity (Metric) 90 mL
Closure Color White
Closure Size 53 mm
Closure Type Bio-Tite Lid
Description Cap and container pre-assembled
Diameter (Metric) 53 mm
Label Tabbed Temp Strip
No. per Case 300/Cs., Pre-assembled
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.