Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMD12 Antibody (OTI2G1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273974AF594
Description
SAMD12 Monoclonal specifically detects SAMD12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SAMD12 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
SAMD12 | |
Full length recombinant protein of human SAMD12 (NP_997389) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2G1 | |
Western Blot, Immunohistochemistry | |
FLJ39458, MGC148139, MGC148140, SAM domain-containing protein 12, sterile alpha motif domain containing 12, sterile alpha motif domain-containing protein 12 | |
Mouse | |
0.1 mL | |
401474 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction