Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMD14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SAMD14 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SAMD14 Polyclonal specifically detects SAMD14 in Human samples. It is validated for Western Blot.Specifications
SAMD14 | |
Polyclonal | |
Rabbit | |
sterile alpha motif domain containing 14 | |
SAMD14 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
201191 | |
Synthetic peptides corresponding to SAMD14(sterile alpha motif domain containing 14) The peptide sequence was selected from the N terminal of SAMD14. Peptide sequence HKARAQLLAKGRRHRPSRSRLRDSASSAEDGEGSDGPGGKVTDGCGSPLH. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title