Learn More
Invitrogen™ SAMD14 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | SAMD14 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SAMD14 |
| Gene Accession No. | Q8IZD0 |
| Gene Alias | AI839049; AI854782; BC034054; RGD1306138; SAM domain-containing protein 14; Samd14; sterile alpha motif domain containing 14; sterile alpha motif domain-containing protein 14 |
| Gene Symbols | SAMD14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.