Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAMD15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556537
Description
Immunogen sequence: RKLIHVNCSN LPQMGITNFE DMKAISRHTQ ELLEIEEPLF KRSISLPYRD IIGLYLEQKG HTGIKSDSLT LSEFVKAAGL QDYAPEITAP E Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 24%.

Specifications
SAMD15 | |
Polyclonal | |
Unconjugated | |
SAMD15 | |
C14orf174; FAM15A; family with sequence similarity 15, member A; Gm263; SAM domain-containing protein 15; SAM domain-containing protein C14orf174; SAMD15; sterile alpha motif domain containing 15; sterile alpha motif domain-containing protein 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
161394 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P1V8 | |
SAMD15 | |
Recombinant protein corresponding to Human SAMD15. Recombinant protein control fragment (Product #RP-95568). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction