Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAMD15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556538 Supplier Invitrogen™ Supplier No. PA556538
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSQEGIAKES KRDVPSETEP GIHQEVKSET SREMGEFFKD LEAPMDETHK ESDLEPPEEA KPNVTEDVFL ESAMETDPDP VP Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 31%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMD15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAMD15
Gene Accession No. Q9P1V8
Gene Alias C14orf174; FAM15A; family with sequence similarity 15, member A; Gm263; SAM domain-containing protein 15; SAM domain-containing protein C14orf174; SAMD15; sterile alpha motif domain containing 15; sterile alpha motif domain-containing protein 15
Gene Symbols SAMD15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAMD15. Recombinant protein control fragment (Product #RP-95569).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161394
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.