Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMD3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA556030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556030 Supplier Thermo Scientific Supplier No. PA556030

Rabbit Polyclonal Antibody

Immunogen sequence: WKRALKDRFK YVRRPIEDDE QVIRNKCKFG HRRGQTRKSL ADIRFDEIKL VQIKEEAVCF DSELDEHIKW FQQEYVKTEK DWR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 60%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMD3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAMD3
Gene Accession No. Q8N6K7
Gene Alias Gm623; SAM domain-containing protein 3; SAMD3; sterile alpha motif domain containing 3; sterile alpha motif domain-containing protein 3; Unknown (protein for MGC:134207)
Gene Symbols SAMD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAMD3
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.