Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAMD4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560224 Supplier Invitrogen™ Supplier No. PA560224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MWLNHLEDRT STSFGGQNRG RSDSVDYGQT HYYHQRQNSD DKLNGWQNSR DSGICINASN WQDKSMGCEN GHVPLYSSSS VPTTINTIGT STS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

Sterile alpha motifs (SAMs) in proteins such as SAMD4A are part of an RNA-binding domain that functions as a posttranscriptional regulator by binding to an RNA sequence motif known as the Smaug recognition element, which was named after the Drosophila Smaug protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMD4A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Samd4a
Gene Accession No. Q9UPU9
Gene Alias 1700024G08Rik; 1700111L17Rik; 4933436G17Rik; KIAA1053; Protein Smaug homolog 1; SAM domain-containing protein 4A; Samd4; SAMD4A; Smaug; smaug 1; Smaug1; SMG; SMGA; sterile alpha motif domain containing 4; sterile alpha motif domain containing 4A; sterile alpha motif domain-containing protein 4A
Gene Symbols Samd4a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAMD4A. Recombinant protein control fragment (Product #RP-100057).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23034
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.