Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMD4B Antibody (PCRP-SAMD4B-1H3), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314152UV
This item is not returnable.
View return policy
Description
SAMD4B Monoclonal antibody specifically detects SAMD4B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
SAMD4B | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
55095 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-SAMD4B-1H3 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
FLJ10211, hSmaug2, MGC99832, protein Smaug homolog 2, SAM domain-containing protein 4B, smaug 2, smaug homolog B, SMAUG2, SMGB, sterile alpha motif domain containing 4B, sterile alpha motif domain-containing protein 4B | |
Recombinant full-length humanSAMD4Bprotein (Uniprot: Q5PRF9) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction