Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMD4B Antibody (PCRP-SAMD4B-1H3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314152MFV610
Description
SAMD4B Monoclonal antibody specifically detects SAMD4B in Human samples. It is validated for Flow CytometrySpecifications
SAMD4B | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
55095 | |
Store at 4C in the dark. | |
IgG2c |
Flow Cytometry | |
PCRP-SAMD4B-1H3 | |
Flow Cytometry | |
FLJ10211, hSmaug2, MGC99832, protein Smaug homolog 2, SAM domain-containing protein 4B, smaug 2, smaug homolog B, SMAUG2, SMGB, sterile alpha motif domain containing 4B, sterile alpha motif domain-containing protein 4B | |
Recombinant full-length humanSAMD4Bprotein (Uniprot: Q5PRF9) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction