Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAMD4B Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550748

Catalog No. PA550748


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total SAMD4B protein.

Has transcriptional repressor activity. Overexpression inhibits the transcriptional activities of AP-1, p53/TP53 and CDKN1A. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SAMD4B
Polyclonal
Unconjugated
SAMD4B
BC042901; hSmaug2; mSmaug2; protein Smaug homolog 2; RGD1305597; SAM domain-containing protein 4B; SAMD4B; Smaug 2; smaug homolog B; Smaug2; SMGB; sterile alpha motif domain containing 4B; sterile alpha motif domain-containing protein 4B
Rabbit
Antigen affinity chromatography
RUO
55095
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q5PRF9
SAMD4B
Synthetic peptide corresponding to a region derived from internal residues of human sterile alpha motif domain containing 4B.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.