Learn More
Invitrogen™ SAMD7 Polyclonal Antibody

Description
SAMD7 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SAMD7 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SAMD7 |
| Gene Accession No. | Q7Z3H4 |
| Gene Alias | 4930597A01Rik; LOW QUALITY PROTEIN: sterile alpha motif domain-containing protein 7; SAM domain-containing protein 7; SAMD7; sterile alpha motif domain containing 7; sterile alpha motif domain-containing protein 7 |
| Gene Symbols | SAMD7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.