Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAMD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554379 Supplier Invitrogen™ Supplier No. PA554379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAKQLNLPEN TDDWTKEDVN QWLESHKIDQ KHREILTEQD VNGAVLKWLK KEHLVDMGIT HGPAIQIEEL FKELRKTAIE DS Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 78%.

Observed MW of SAMD9 is from 148 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMD9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAMD9
Gene Accession No. Q5K651
Gene Alias C7orf5; DRIF1; expressed in aggressive fibromatosis; KIAA2004; NFTC; OEF1; OEF2; RGD1563091; SAM domain-containing protein 9; SAMD9; similar to OEF2; sterile alpha motif domain containing 9; sterile alpha motif domain-containing protein 9
Gene Symbols SAMD9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAMD9. Recombinant protein control fragment (Product #RP-92853).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.