Learn More
Invitrogen™ SAMD9 Recombinant Rabbit Monoclonal Antibody (K01_1Y09)

Rabbit Recombinant Monoclonal Antibody
$201.75 - $389.25
Specifications
| Antigen | SAMD9 |
|---|---|
| Clone | K01_1Y09 |
| Concentration | 1 mg/mL |
| Content And Storage | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
| Applications | Western Blot |
Description
Immunogen sequence: QTSKMGKPSK NAPKDQTVSQ KERRETSKQK QKGKENPDMA NPSAMSTTAK GSKSLKVELI EDKIDYTKER QPSIDLTCVS YPFDEFSNPY RYKLDFSLQP ETGPGN Highest antigen sequence identity to the following orthologs: Pig - 57%, Cynomolgus monkey - 92%.
Observed MW of SAMD9 is from 148 kDa.Specifications
| SAMD9 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 | |
| Q5K651 | |
| 54809 | |
| A synthetic peptide of human SAMD9 (aa 84-189). | |
| Antibody |
| K01_1Y09 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| SAMD9 | |
| C7orf5; DRIF1; expressed in aggressive fibromatosis; KIAA2004; NFTC; OEF1; OEF2; RGD1563091; SAM domain-containing protein 9; SAMD9; similar to OEF2; sterile alpha motif domain containing 9; sterile alpha motif domain-containing protein 9 | |
| SAMD9 | |
| Primary | |
| Protein A/G |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.